Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PP2C epsilon/PPM1L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6914SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6914SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6914SP Supplier R&D Systems Supplier No. MAB6914SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PP2C epsilon/PPM1L Monoclonal specifically detects PP2C epsilon/PPM1L in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PP2C epsilon/PPM1L
Applications Western Blot
Classification Monoclonal
Clone 700612
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q5SGD2
Gene Alias EC 3.1.3, EC 3.1.3.16, MGC132545, MGC132547, PP2C epsilon, PP2CE, PP2CEProtein phosphatase 2C isoform epsilon, PP2C-epsilon, PPM1L, PPM1-LIKE, protein phosphatase 1 (formerly 2C)-like, protein phosphatase 1L, Protein phosphatase 1-like, Protein phosphatase 2C epsilon isoform, protein phosphatase, Mg2+/Mn2+ dependent, 1L
Gene Symbols PPM1L
Host Species Mouse
Immunogen E. coli-derived recombinant human PP2C epsilon/PPPM1L Asp43-Arg178 Accession # Q5SGD2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151742
Test Specificity Detects human PP2C epsilon/PPPM1L in direct ELISAS and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human PP2C is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.