Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PPP2R4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5929
This item is not returnable.
View return policy
Description
PPP2R4 Monoclonal specifically detects PPP2R4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PPP2R4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 5.2.1.8, MGC2184, Phosphotyrosyl phosphatase activator, PP2A, PP2A phosphatase activator, PP2A subunit B' isoform PR53, PP2A, subunit B', PR53 isoform, protein phosphatase 2A activator, regulatory subunit 4, PTPA, regulatory subunit B' (PR 53), serine/threonine-protein phosphatase 2A activator, Serine/threonine-protein phosphatase 2A regulatory subunit 4, Serine/threonine-protein phosphatase 2A regulatory subunit B' | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5524 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot | |
529245 | |
Western Blot 0.5 ug/mL | |
Q15257 | |
PPP2R4 | |
E. coli-derived recombinant human PPP2R4 Glu14-Asn323 Accession # Q15257 | |
100 μg | |
Primary | |
Detects endogenous human, mouse and rat PPP2R4 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction