Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat PTP-MEG2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2668SP
This item is not returnable.
View return policy
Description
PTP-MEG2/PTPN9 Polyclonal specifically detects PTP-MEG2/PTPN9 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PTP-MEG2/PTPN9 | |
Polyclonal | |
Western Blot 1 ug/mL | |
P43378 | |
PTPN9 | |
E. coli-derived recombinant human PTP-MEG2 Met1-Gln593 Accession # P43378 | |
25 μg | |
Primary | |
Detects human, mouse, and rat PTP-MEG2 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.3.48, MEG2, protein tyrosine phosphatase, non-receptor type 9, Protein-tyrosine phosphatase MEG2, PTPase MEG2, PTPase-MEG2, PTPMEG2, PTPN9, tyrosine-protein phosphatase non-receptor type 9 | |
Goat | |
Affinity Purified | |
RUO | |
5780 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction