Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat PTP mu/PTPRM Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4446
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4446 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4446 Supplier R&D Systems Supplier No. MAB4446
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTP mu/PTPRM Monoclonal specifically detects PTP mu/PTPRM in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PTP mu/PTPRM
Applications Western Blot
Classification Monoclonal
Clone 436502
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P28827
Gene Alias EC 3.1.3.48, hR-PTPu, MGC166994, protein tyrosine phosphatase mu, protein tyrosine phosphatase, receptor type, M, protein tyrosine phosphatase, receptor type, mu polypeptide, Protein-tyrosine phosphatase mu, PTPRL1, PTPRL1R-PTP-MU, PTPRM, receptor-type tyrosine-protein phosphatase mu, RPTPM, R-PTP-mu, RPTPU
Gene Symbols PTPRM
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PTPμ/PTPRM Glu21-Lys742 Accession # P28827
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5797
Test Specificity Detects human, mouse, and rat PTPμ/PTPRM in Western blots. In Western blots, no cross-reactivity with recombinant human PTPRK, PTPRT, PTPRD, PTPRG, PTPRF (LAR), or DEP1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.