Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat RAGE Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1179SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1179SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1179SP Supplier R&D Systems Supplier No. AF1179SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 11 publications

AGER Polyclonal specifically detects AGER in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.

Specifications

Antigen RAGE
Applications Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1-1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O35444
Gene Alias advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1
Gene Symbols Ager
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gln24-Ala342 Accession # O35444
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5891
Test Specificity Detects human, mouse, and rat RAGE in Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant canine RAGE is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.