Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat RalA Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3270SP
This item is not returnable.
View return policy
Description
RalA Polyclonal specifically detects RalA in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RalA | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HINCONS, RAL, Rasl1 | |
Goat | |
Affinity Purified | |
RUO | |
5898 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
P11233 | |
RALA | |
E. coli-derived recombinant human RalA Met1-Leu206 Accession # P11233 | |
25 μg | |
Primary | |
Detects human, mouse and rat RalA in Western blots. When immunoblotted, no cross-reactivity with recombinant human (rh) RalB, rhH-Ras, rhN-Ras or rhK-Ras is observed. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction