Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat RalA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3270
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3270 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3270 Supplier R&D Systems Supplier No. MAB3270
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

RalA Monoclonal specifically detects RalA in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen RalA
Applications Western Blot
Classification Monoclonal
Clone 399527
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P11233
Gene Alias HINCONS, RAL, Rasl1
Gene Symbols RALA
Host Species Rat
Immunogen E. coli-derived recombinant human RalA Met1-Leu206 Accession # P11233
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5898
Test Specificity Detects human, mouse, and rat RalA in Western blots. In Western blots, no cross-reactivity with recombinant human RalB is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.