Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat RalA/RalB Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3204
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3204 Supplier R&D Systems Supplier No. AF3204
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

RalA/RalB Polyclonal specifically detects RalA/RalB in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen RalA/RalB
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P11234
Gene Alias MGC48949, RAL, Ras family small GTP binding protein RALA, Rasl1, RAS-like protein A, ras-related protein Ral-A, v-ral simian leukemia viral oncogene homolog A (ras related)
Gene Symbols RALA
Host Species Goat
Immunogen E. coli-derived recombinant human RalB Met1-Leu206 Accession # P11234
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5898
Test Specificity Detects human, mouse, and rat RalA/RalB in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human H-Ras, N-Ras, or K-Ras is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.