Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat RBBP4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7416SP
This item is not returnable.
View return policy
Description
RBBP4/RbAp48 Monoclonal specifically detects RBBP4/RbAp48 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RBBP4/RbAp48 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CAF-1 p48, CAF-1 subunit C, CAF-1C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48 | |
E. coli-derived recombinant human RBBP-4 Ala2-Ser425 Accession # 09028 | |
25 μg | |
Primary | |
Detects human RBBP-4 in direct ELISAs and Western blots. | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
968701 | |
Western Blot 0.5 ug/mL, Immunohistochemistry 5 ug/mL | |
9028 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5928 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction