Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat RBBP4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7416
This item is not returnable.
View return policy
Description
RBBP4/RbAp48 Polyclonal specifically detects RBBP4/RbAp48 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RBBP4/RbAp48 | |
Polyclonal | |
Unconjugated | |
Q09028 | |
RBBP4 | |
E. coli-derived recombinant human RBBP4 Ala2-Ser425 Accession # Q09028 | |
100 μg | |
Primary | |
Detects human, mouse and rat RBBP4 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human RBBP7 is observed. Detects recombinant human RBBP4 in direct ELISAs. In direct ELISAs, approximately 75% cross-reactivity with recombinant human RBBP7 is observed. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
CAF-1 p48, CAF-1 subunit C, CAF-1C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48 | |
Sheep | |
Affinity Purified | |
RUO | |
5928 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction