Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat RelA/NF kappa B p65 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF5078SP
This item is not returnable.
View return policy
Description
RelA/NFkB p65 Polyclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Knockout Validated.Specifications
RelA/NFkB p65 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, p65RelA, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
Sheep | |
Affinity Purified | |
RUO | |
5970 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
ChIP Assay, Immunocytochemistry, Immunohistochemistry | |
LYOPH | |
Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 5-15 μg/mL, Chromatin Immunoprecipitation (ChIP) 5 μg/5 x 10^6 cells, Immunocytochemistry 5-15 μg/mL, Knockout Validated 1 μg/mL | |
Q04206 | |
RELA | |
E. coli-derived recombinant human RelA/NF kappa B p65 isoform 1 Asn456-Ser551 Accession # Q04206 | |
25 μg | |
Primary | |
Detects human and mouse RelA/NF kappa B p65 in Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction