Learn More
Human/Mouse/Rat Relaxin R1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Relaxin R1/LGR7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 933344 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q9HBX9 |
| Gene Alias | leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RelaxinR1, RXFP1, RXFPR1 |
| Host Species | Mouse |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.