Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Relaxin R1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$222.00 - $558.00
Specifications
Antigen | Relaxin R1/LGR7 |
---|---|
Clone | 933344 |
Dilution | Western Blot 2 μg/mL, Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 1-25 μg/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Description
Relaxin R1/LGR7 Monoclonal specifically detects Relaxin R1/LGR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
Relaxin R1/LGR7 | |
Western Blot 2 μg/mL, Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 1-25 μg/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RelaxinR1, RXFP1, RXFPR1 | |
RXFP1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
933344 | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q9HBX9 | |
59350 | |
Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R1 Met1-Ser398 Accession # Q9HBX9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Relaxin R1 in direct ELISAs and detects human, mouse, and rat Relaxin R1 in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title