Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Ribosomal Protein S6/RPS6 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC5436G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5436G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5436G Supplier R&D Systems Supplier No. IC5436G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ribosomal Protein S6/RPS6 Monoclonal specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry.

Specifications

Antigen Ribosomal Protein S6/RPS6
Applications Immunocytochemistry
Classification Monoclonal
Clone 522731
Conjugate Alexa Fluor 488
Dilution Immunocytochemistry 1:10 dilution
Gene Accession No. P62753
Gene Alias Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, RPS6, S6
Gene Symbols RPS6
Host Species Mouse
Immunogen E. coli-derived recombinant human Ribosomal Protein S6/RPS6 Met1-Lys249 Accession # P62753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 6194
Test Specificity Conjugated Ribosomal Protein S6/RPS6 antibodies are ideal for immunocytochemistry colocalization studies in ribosomes. The unconjugated antibody detects endogenous human, mouse and rat Ribosomal Protein S6/RPS6 in Western blots.
Target Species Human, Mouse, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.