Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat SHIP2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5389
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5389 Supplier R&D Systems Supplier No. AF5389
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

SHIP2/INPPL1 Polyclonal specifically detects SHIP2/INPPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen SHIP2/INPPL1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. O15357
Gene Alias EC 3.1.3, EC 3.1.3.n1, inositol polyphosphate phosphatase-like 1, Inositol polyphosphate phosphatase-like protein 1, INPPL1, INPPL-1, phosphatidylinositol-34,5-trisphosphate 5-phosphatase 2, Protein 51C, SH2 domain-containing inositol phosphatase 2, SH2 domain-containing inositol-5'-phosphatase 2, SHIP-2, SHIP251C protein
Gene Symbols INPPL1
Host Species Sheep
Immunogen E. coli-derived recombinant human SHIP2 Ala1106-Lys1258 Accession # O15357
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3636
Test Specificity Detects human, mouse and rat SHIP2 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.