Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat SHP-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1894
This item is not returnable.
View return policy
Description
SHP-2/PTPN11 Monoclonal specifically detects SHP-2/PTPN11 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
SHP-2/PTPN11 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
NP_002825 | |
PTPN11 | |
E. coli-derived recombinant human SHP-2 Thr2-Arg593 Accession # NP_002825 | |
200 μg | |
Primary | |
Detects human, mouse, and rat SHP-2 in Western blots. In Western blots, no cross-reactivity with recombinant human SHP-1 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
255509 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP1D, PTP-1D, PTP2C, PTP-2C, PTP2CNS1, PTPN11, SHP2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5781 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction