Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat SHP-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1894SP
This item is not returnable.
View return policy
Description
SHP-2/PTPN11 Monoclonal specifically detects SHP-2/PTPN11 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
SHP-2/PTPN11 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
NP_002825 | |
PTPN11 | |
E. coli-derived recombinant human SHP-2 Thr2-Arg593 Accession # NP_002825 | |
25 μg | |
Primary | |
Detects human, mouse, and rat SHP-2 in Western blots. In Western blots, no cross-reactivity with recombinant human SHP-1 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
255509 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP1D, PTP-1D, PTP2C, PTP-2C, PTP2CNS1, PTPN11, SHP2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5781 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction