Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Slit3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3629
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3629 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3629 Supplier R&D Systems Supplier No. AF3629
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Slit3 Polyclonal specifically detects Slit3 in Human, Mouse, Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Slit3
Applications Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5 μg/mL, Neutralization
Gene Accession No. Q9WVB4
Gene Alias KIAA0814, MEGF5, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3slit (Drosophila) homolog 3
Gene Symbols SLIT3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Slit3 Ser27-His901 Accession # Q9WVB4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6586
Test Specificity Detects human, mouse and rat Slit3 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.