Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Sortilin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3154SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3154SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3154SP Supplier R&D Systems Supplier No. MAB3154SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sortilin Monoclonal specifically detects Sortilin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Knockout Validated.

Specifications

Antigen Sortilin
Applications Western Blot, KnockDown
Classification Monoclonal
Clone 334708
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25-1 ug/mL, Simple Western 10 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99523
Gene Alias 100 kDa NT receptor, Glycoprotein 95, Gp95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, Ntr3, SORT1, sortilin, sortilin 1
Gene Symbols SORT1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Sortilin Ser78-Asn755 Accession # Q99523
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6272
Test Specificity Detects human, mouse, and rat Sortilin in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.