Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat SOX2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2018R100UG
This item is not returnable.
View return policy
Description
SOX2 Monoclonal specifically detects SOX2 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
SOX2 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P48431 | |
SOX2 | |
E. coli-derived recombinant human SOX2 Gly135-Met317 Accession # P48431 | |
RUO | |
6657 | |
Human, Mouse, Rat | |
IgG2a |
Flow Cytometry | |
245610 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
Mouse | |
100 μg | |
Primary | |
Detects human, mouse, and rat SOX2 in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction