Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat SOX2 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF2018 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2018 Supplier R&D Systems Supplier No. AF2018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 119 publications

SOX2 Polyclonal specifically detects SOX2 in Aedes albopictus (Asian tiger mosquito)∣Bovine∣Callithrix jacchus (Common Marmoset)∣Canine∣Chicken∣Cynomolgus Monkey∣Ferret∣Fish∣Human∣Mouse∣Pan troglodytes (Chimpanzee)∣Pig∣Quail∣Rabbit∣Rat∣Rhesus Macaque∣Transgenic Mouse∣Xenograft∣Xenopus species∣Zebrafish samples. It is validated for Binding Assay∣Bioassay∣ChIP Assay∣Co-immunoprecipitation (Co-IP)∣Dot Blot∣Flow Cytometry∣IF/ICC∣Immunocytochemistry∣Immunocytochemistry/Immunofluorescence∣Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣Mass Spectrometry∣RNAscope compatible∣Simple Western∣Western Blot.

Specifications

Antigen SOX2
Applications Binding Activity, Bioassay, ChIP Assay, Co-immunoprecipitation (Co-IP), Dot Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Mass Spectrometry, Dual RNAScope ISH-IHC, Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 3-25 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P48431
Gene Alias ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Gene Symbols SOX2
Host Species Goat
Immunogen E. coli-derived recombinant human SOX2 Gly135-Met317 Accession # P48431
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6657
Test Specificity Detects human SOX2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Asian Tiger Mosquito, Bovine, Marmoset, Canine, Chicken, Cynomolgus Monkey, Ferret, Fish, Human, Mouse, Chimpanzee, Pig, Quail, Rabbit, Rat, Rhesus Macaque, Transgenic Mouse, Xenograft, Xenopus species, Zebrafish
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.