Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat SP-D Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1920
This item is not returnable.
View return policy
Description
SP-D Polyclonal specifically detects SP-D in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
SP-D | |
Polyclonal | |
Unconjugated | |
P35247 | |
SFTPD | |
Mouse myeloma cell line NS0-derived recombinant human SP-D Ala21-Phe375 (Glu22Gly) Accession # P35247 | |
100 μg | |
Primary | |
Detects human SP-D in direct ELISAs and human, mouse, and rat SP-D in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL, Simple Western 10 ug/mL | |
COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4, SFTP4pulmonary surfactant-associated protein D, SFTPD, SPD, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
Goat | |
Affinity Purified | |
RUO | |
6441 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction