Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7275
This item is not returnable.
View return policy
Description
ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Polyclonal specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 | |
Polyclonal | |
Unconjugated | |
NP_008858 | |
ST3GAL2 | |
Chinese hamster ovary cell line CHO-derived recombinant human ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Pro52-Gln350 Accession # NP_008858 | |
100 μg | |
Primary | |
Detects human, mouse and rat ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ST6GALNAC4 and rhST3GAL1 is observed. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
Alpha 2,3-ST 2, Beta-galactoside alpha-2,3-sialyltransferase 2, beta-galactoside alpha-2,3-sialytransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2, EC 2.4.99.4, EC 2.4.99.7, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4B, sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase), SIAT4B, SIAT4-B, ST3 beta-galactoside alpha-2,3-sialyltransferase 2, ST3GalA.2, ST3GalA.2ST3Gal II, ST3GALII | |
Sheep | |
Affinity Purified | |
RUO | |
6483 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction