Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat STAT3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1799
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1799 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1799 Supplier R&D Systems Supplier No. MAB1799
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

STAT3 Monoclonal specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated.

Specifications

Antigen STAT3
Applications Western Blot, Immunoprecipitation, Flow Cytometry, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 232209
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 1-3 ug/500 ug cell lysate, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P40763
Gene Alias Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Gene Symbols STAT3
Host Species Mouse
Immunogen E. coli-derived recombinant human STAT3 Met1-Asn175 Accession # P40763
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6774
Test Specificity Detects human, mouse, and rat STAT3 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.