Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat T Cell Receptor alpha Chain-V alpha 24-J alpha Q Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4988
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4988 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4988 Supplier R&D Systems Supplier No. AF4988
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

T Cell Receptor alpha Chain-V alpha 24-J alpha Q Polyclonal specifically detects T Cell Receptor alpha Chain-V alpha 24-J alpha Q in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen T Cell Receptor alpha Chain-V alpha 24-J alpha Q
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. ABC72391
Host Species Sheep
Immunogen E. coli-derived recombinant human T Cell Receptor alpha Chain-V alpha 24-J alpha Q Lys22-Pro135 Accession # ABC72391
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human, mouse, and rat T Cell Receptor alpha Chain-V alpha 24-J alpha Q in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.