Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Talin1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5456
This item is not returnable.
View return policy
Description
Talin1 Polyclonal specifically detects Talin1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Talin1 | |
Polyclonal | |
Unconjugated | |
Q9Y490 | |
TLN1 | |
E. coli-derived recombinant human Talin1 Ala975-Ser1181 Accession # Q9Y490 | |
100 μg | |
Primary | |
Detects human, mouse and rat Talin1 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
ILWEQ, KIAA1027, talin 1, talin-1, TLN, TLN1 | |
Goat | |
Affinity Purified | |
RUO | |
7094 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction