Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat TC-PTP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1930
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1930 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1930 Supplier R&D Systems Supplier No. AF1930
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TC-PTP/PTPN2 Polyclonal specifically detects TC-PTP/PTPN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen TC-PTP/PTPN2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL
Gene Accession No. P17706
Gene Alias protein tyrosine phosphatase, non-receptor type 2, PTP2, PTPN2, PTPTPTN2, T-cell protein tyrosine phosphatase, T-cell protein-tyrosine phosphatase, TCELLPTP, TCPTP, TC-PTP, TCPTPEC 3.1.3.48, tyrosine-protein phosphatase non-receptor type 2
Gene Symbols PTPN2
Host Species Goat
Immunogen E. coli-derived recombinant human TC-PTP Pro2-Asn314 Accession # P17706
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5771
Test Specificity Detects human, mouse and rat TC-PTP in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.