Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat TDP-43/TARDBP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB77781SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB77781SP 25 μg
MAB77781100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB77781SP Supplier R&D Systems Supplier No. MAB77781SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TDP-43/TARDBP Monoclonal specifically detects TDP-43/TARDBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen TDP-43/TARDBP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 982022
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q13148
Gene Alias ALS10, ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TARDBP, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Host Species Mouse
Immunogen E.coli-derived recombinant human TDP-43/TARDBP Met1-Thr103 Accession # Q13148
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 23435
Test Specificity Detects human TDP-43/TARDBP in direct ELISAs. Detects human, mouse, and rat TDP-43/TARDBP in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.