Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Testican 2/SPOCK2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2328SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2328SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2328SP Supplier R&D Systems Supplier No. AF2328SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Testican 2/SPOCK2 Polyclonal specifically detects Testican 2/SPOCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Testican 2/SPOCK2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q92563
Gene Alias FLJ97039, KIAA0275, KIAA0275CWCV, and Kazal-like domains proteoglycan 2, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2, SPOCK2
Gene Symbols SPOCK2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Testican 2/SPOCK2 Glu23-Trp424 Accession # Q92563
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9806
Test Specificity Detects human, mouse, and rat Testican 2/SPOCK2 in direct ELISAs and human Testican 2/SPOCK2 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.