Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Thioredoxin Reductase 1/TRXR1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC7428N100UG
This item is not returnable.
View return policy
Description
Thioredoxin Reductase 1/TRXR1 Monoclonal specifically detects Thioredoxin Reductase 1/TRXR1 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Thioredoxin Reductase 1/TRXR1 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q16881 | |
TXNRD1 | |
E. coli-derived recombinant mouse Thioredoxin Reductase 1/TRXR1 Met1-Ile497 Accession # Q16881 | |
RUO | |
7296 | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
489804 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 1.8.1, EC 1.8.1.9, Gene associated with retinoic and IFN-induced mortality 12 protein, Gene associated with retinoic and interferon-induced mortality 12 protein, GRIM12, GRIM-12, GRIM-12TR1, KDRF, KM-102-derived reductase-like factor, MGC9145, oxidoreductase, thioredoxin reductase 1, thioredoxin reductase 1, cytoplasmic, thioredoxin reductase GRIM-12, Thioredoxin reductase TR1, TR, TR1, Trxr1, TRXRD1, TXNR, TXNRD1, TXNRTRXR1 | |
Mouse | |
100 μg | |
Primary | |
Detects mouse Thioredoxin Reductase 1/TRXR1 in direct ELISAs and human, mouse, and rat Thioredoxin Reductase 1/TRXR1 in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction