Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Thymosin beta 10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6429
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6429 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6429 Supplier R&D Systems Supplier No. AF6429
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Thymosin beta 10 Polyclonal specifically detects Thymosin beta 10 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Thymosin beta 10
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P63313
Gene Alias MIG12, migration-inducing gene 12, migration-inducing protein 12, PTMB10, TB10, THYB10, thymosin beta 10, thymosin beta-10, TMSB10
Gene Symbols TMSB10
Host Species Sheep
Immunogen E. coli-derived recombinant human Thymosin beta 10 Ala2-Ser44 Accession # P63313
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9168
Test Specificity Detects human Thymosin beta 10 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Thymosin beta 4 and rhThymosin beta 16 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.