Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Tmp21/p23 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7630100
This item is not returnable.
View return policy
Description
Tmp21/p23 Monoclonal specifically detects Tmp21/p23 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
Tmp21/p23 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMED10, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein | |
E.coli-derived recombinant human Tmp21/p23 Ile32-Arg185 Accession # P49755 | |
100 μg | |
Primary | |
Detects human Tmp21/p23 in direct ELISAs and Western blots. | |
Human, Mouse, Rat | |
IgG2a |
Western Blot | |
805125 | |
Western Blot 1 ug/mL | |
P49755 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10972 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction