Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat Total Akt1 DuoSet IC ELISA

Catalog No. DYC17752
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC17752 2 Plates
DYC1775E 15 Plates
DYC17755 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC17752 Supplier R&D Systems™ Supplier No. DYC17752
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human/Mouse/Rat Total Akt1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human/Mouse/Rat Total Akt1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat AKT1.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit (for 2 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 312.00 to 20000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 207
Synonym AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-Pk.-alpha, RACPKB-ALPHA, Serine-Threonine Protein Kinase, v-akt murine thymoma viral oncogene homolog 1, V-Akt Murine Thymoma Viral Oncogene-Like Protein 1
Packaging Quantity 2 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.