Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat Total HSP70/HSPA1A DuoSet IC ELISA

Catalog No. DYC16632
Change view
Click to view available options
Format:
15 96-well Plates
2 96-well Plates
5 96-well Plates
Gene ID (Entrez):
3303
24472
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Gene ID (Entrez)
DYC16632 2 96-well Plates 24472
DYC1663E 15 96-well Plates 3303
DYC16635 5 96-well Plates 24472
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC16632 Supplier R&D Systems™ Supplier No. DYC16632
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human/Mouse/Rat Total HSP70/HSPA1A DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human/Mouse/Rat Total HSP70/HSPA1A DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat HSP70/HSPA1A.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 156.00 to 10000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 2 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 24472
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.