Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat TPT1/TCTP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4648SP
This item is not returnable.
View return policy
Description
TPT1/TCTP Monoclonal specifically detects TPT1/TCTP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
TPT1/TCTP | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL | |
P13693 | |
TPT1 | |
E. coli-derived recombinant human TPT1/TCTP Met1-Cys172 Accession # P13693 | |
25 μg | |
Primary | |
Detects human, mouse, and rat TPT1/TCTP in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry | |
488411 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ27337, Fortilin, Histamine-releasing factor, HRF, p02, p23, TCTP, TCTPHRFfortilin, translationally-controlled tumor protein, tumor protein, translationally-controlled 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7178 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction