Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat TrkB Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1494
This item is not returnable.
View return policy
Description
TrkB Monoclonal specifically detects TrkB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TrkB | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P15209 | |
NTRK2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TrkB Cys32-His429 Accession # P15209 | |
500 μg | |
Primary | |
Detects mouse TrkB in direct ELISAs. Detects human, mouse, and rat TrkB in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TrkA, recombinant rat TrkA, or recombinant mouse TrkC is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
225105 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, NTRK2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4915 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction