Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat UCH-L1/PGP9.5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB60072SP
This item is not returnable.
View return policy
Description
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Specifications
UCH-L1/PGP9.5 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7345 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
972119 | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 0.3-25 ug/mL | |
P09936 | |
UCHL1 | |
E. coli-derived recombinant human UCH-L1/PGP9.5 Gln2-Ala223 Accession # P09936 | |
25 μg | |
Primary | |
Detects human UCH-L1/PGP9.5 in direct ELISAs. Detects human, mouse, and rat in Western blot. | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction