Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat VHR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1654SP
This item is not returnable.
View return policy
Description
VHR/DUSP3 Monoclonal specifically detects VHR/DUSP3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VHR/DUSP3 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P51452 | |
DUSP3 | |
E. coli-derived recombinant human VHR Ser2-Pro185 Accession # P51452 | |
25 μg | |
Primary | |
Detects human, mouse, and rat VHR in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
237020 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dual specificity phosphatase 3, dual specificity protein phosphatase 3, Dual specificity protein phosphatase VHR, DUSP3, EC 3.1.3.16, EC 3.1.3.48, Vaccinia H1-related phosphatase, vaccinia virus phosphatase VH1-related, VHRserine/threonine specific protein phosphatase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1845 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction