Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Vimentin Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC2105U100UG
This item is not returnable.
View return policy
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| Vimentin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| FLJ36605, VIM, vimentin | |
| E. coli-derived recombinant human Vimentin, Ser2-Glu466, Accession No. P08670 | |
| 100 μg | |
| 7431.0 | |
| Human, Mouse, Rat | |
| Supernatant |
| Flow Cytometry | |
| 280618 | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| P08670 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Vimentin antibodies are ideal for immunocytochemistry colocalization studies in intermediate filaments. Detects human, mouse and rat Vimentin in Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction