Learn More
Human/Mouse/Rat Vimentin Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Vimentin |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated 5-15 ug/mL |
| Gene Accession No. | P08670 |
| Gene Alias | FLJ36605, VIM, vimentin |
| Gene Symbols | VIM |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.