Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human, Mouse, Rat VPS35 ELISA Kit (Colorimetric)
SDP

Catalog No. NB397987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 wells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397987 96 wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397987 Supplier Novus Biologicals™ Supplier No. NBP311045
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 78.125 to 5000 pg/mL
Assay Sensitivity 50 pg/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Lysates, Tissue Extracts
Target Species Human, Mouse, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 55737
Gene Symbol VPS35
Quantity 96 wells
Research Discipline Autophagy, Cellular Markers
Storage Requirements Store at 4°C.
Synonym DKFZp434P1672, FLJ10752, FLJ13588, FLJ20388, hVPS35, Maternal-embryonic 3, MEM3DKFZp434E1211, vacuolar protein sorting 35 (yeast homolog), vacuolar protein sorting 35 (yeast), vacuolar protein sorting 35 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 35, Vesicle protein sorting 35
Target Human, Mouse, Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.