Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat XIAP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8221SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8221SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8221SP Supplier R&D Systems Supplier No. AF8221SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 14 publications

XIAP Polyclonal specifically detects XIAP in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.

Specifications

Antigen XIAP
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P98170
Gene Alias API3X-linked inhibitor of apoptosis protein, apoptosis inhibitor 3, baculoviral IAP repeat-containing 4, baculoviral IAP repeat-containing protein 4, BIRC4, BIRC4XLP2, E3 ubiquitin-protein ligase XIAP, EC 6.3.2.-, hIAP3, hIAP-3, hILP, IAP3, IAP-3, IAP-like protein, ILP, ILP1, Inhibitor of apoptosis protein 3, MIHA, X-linked IAP, X-linked inhibitor of apoptosis
Gene Symbols XIAP
Host Species Goat
Immunogen E. coli-derived recombinant human XIAP Met1-Ser497 (Ser162Cys) Accession # P98170
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 331
Test Specificity Detects human, mouse and rat XIAP in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.