Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat XPD Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3296
This item is not returnable.
View return policy
Description
XPD Polyclonal specifically detects XPD in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
XPD | |
Polyclonal | |
Unconjugated | |
P18074 | |
ERCC2 | |
E. coli-derived recombinant human XPD Met1-Gly360 Accession # P18074 | |
100 μg | |
Primary | |
Detects human, mouse, and rat XPD in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
Basic transcription factor 2 80 kDa subunit, BTF2 p80, COFS2, CXPD, DNA excision repair protein ERCC-2, DNA repair protein complementing XP-D cells, EC 3.6.1, EC 3.6.4.12, EM9, EM9TFIIH basal transcription factor complex 80 kDa subunit, ERCC2, excision repair cross-complementing rodent repair deficiency, complementationgroup 2, MAG, MGC102762, MGC126218, MGC126219, TFIIH 80 kDa subunit, TFIIH basal transcription factor complex helicase subunit, TFIIH basal transcription factor complex helicase XPD subunit, TTD, xeroderma pigmentosum complementary group D, Xeroderma pigmentosum group D-complementing protein, XPDC, XPDTFIIH p80 | |
Goat | |
Affinity Purified | |
RUO | |
2068 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction