Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse RelA/NFkB p65 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC5078T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5078T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5078T100 Supplier R&D Systems Supplier No. IC5078T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen RelA/NFkB p65
Applications Flow Cytometry
Classification Monoclonal
Clone 532301
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q04206
Gene Alias MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, p65RelA, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Gene Symbols RELA
Host Species Mouse
Immunogen E. coli-derived recombinant human RelA/NF kappa B p65 isoform 1 Asn456-Ser551 Accession # Q04206
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5970
Test Specificity Detects human and mouse RelA/NF kappa B p65 in Western blots.
Target Species Human, Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.