Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Ribosomal Protein L17 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7158
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7158 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7158 Supplier R&D Systems Supplier No. MAB7158
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ribosomal Protein L17 Monoclonal specifically detects Ribosomal Protein L17 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Ribosomal Protein L17
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 702142
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P18621
Gene Alias FLJ18762,60S ribosomal protein L23, FLJ92089, gene encoding putative NFkB activating protein, MGC117162, PD-1, ribosomal protein L17, RPL17, RPL23, RPL23,60S ribosomal protein L17
Gene Symbols RPL17
Host Species Mouse
Immunogen E. coli-derived recombinant human Ribosomal Protein L17 Cys70-Glu184 Accession # P18621
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6139
Test Specificity Detects human Ribosomal Protein L17 in direct ELISAs, and human and mouse Ribosomal Protein L17 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.