Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse RNF168 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7217
This item is not returnable.
View return policy
Description
RNF168 Polyclonal specifically detects RNF168 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
RNF168 | |
Polyclonal | |
Unconjugated | |
Q80XJ2 | |
RNF168 | |
E. coli-derived recombinant mouse RNF168 Asn423-Arg565 Accession # Q80XJ2 | |
100 μg | |
Primary | |
Detects recombinant mouse RNF168 and recombinant human RNF168 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
E3 ubiquitin-protein ligase RNF168, EC 6.3.2, EC 6.3.2.-, FLJ39749, ring finger protein 168FLJ35794 | |
Sheep | |
Affinity Purified | |
RUO | |
165918 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction