Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse ROR gamma t/RORC2/NR1F3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC9125G025
This item is not returnable.
View return policy
Description
ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| ROR gamma t/RORC2/NR1F3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Rorc | |
| E.coli-derived recombinant human ROR gamma t/RORC2/NR1F3 Met1-Gln100 Accession # P51449 | |
| 25 Tests | |
| Primary | |
| Detects recombinant human ROR gamma t/RORC2/NR1F3 by direct ELISAs and detects human and mouse ROR gamma t/RORC2/NR1F3 in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
| Flow Cytometry | |
| 1181A | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| P51449 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6097 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction