Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse ROR gamma t/RORC2/NR1F3 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC6006A025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6006A025 25 Tests
IC6006A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. IC6006A025 Supplier R&D Systems Supplier No. IC6006A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen ROR gamma t/RORC2/NR1F3
Applications Flow Cytometry
Classification Monoclonal
Clone 600380
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P51449
Gene Symbols RORC
Host Species Mouse
Immunogen Human ROR gamma t peptide Met1-Arg10 Accession # P51449
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6097
Test Specificity Detects human and mouse ROR gamma t/RORC2 in direct ELISAs. In direct ELISAs, no cross-reactivity with human and mouse RORC.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.