Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse ROR gamma t/RORC2/NR1F3 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC9125A025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC9125A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC9125A025 Supplier R&D Systems Supplier No. IC9125A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen ROR gamma t/RORC2/NR1F3
Applications Flow Cytometry
Classification Monoclonal
Clone 1181A
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P51449
Gene Symbols Rorc
Host Species Rabbit
Immunogen E. coli derived recombinant human ROR gamma T/RORC2/NR1F3, Met1-Gln100, Accession # P51449
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6097
Test Specificity Detects recombinant human ROR gamma t/RORC2/NR1F3 by direct ELISAs and detects human and mouse ROR gamma t/RORC2/NR1F3 in flow cytometry.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.