Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse ROR gamma t/RORC2/NR1F3 PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 1 publication

$268.50 - $503.00

Specifications

Antigen ROR gamma t/RORC2/NR1F3
Clone 600380
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
IC6006C025
GREENER_CHOICE
View Documents
R&D Systems
IC6006C025
25 Tests
Each for $268.50
Only null left
Add to Cart
 
IC6006C100
GREENER_CHOICE
View Documents
R&D Systems
IC6006C100
100 Tests
Each for $503.00
Only null left
Add to Cart
 
Description

Description

ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications

Specifications

ROR gamma t/RORC2/NR1F3
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
6097
Human ROR gamma t peptide Met1-Arg10 Accession # P51449
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
600380
Flow Cytometry
PerCP
Mouse
Human, Mouse
P51449
RORC
IgG2b
Protein A or G purified from hybridoma culture supernatant
Detects human and mouse ROR gamma t/RORC2 in direct ELISAs. In direct ELISAs, no cross-reactivity with human and mouse RORC.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.